Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ING4 Antibodies

          Rockland

          p29ING4 Polyclonal Antibody

          View all (12) ING4 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite p29ING4 Polyclonal Antibody

          • Antibody Testing Data (2)
          p29ING4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          p29ING4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          p29ING4 Antibody (600-101-284) in WB

          Anti-p29 ING4 polyclonal antibody detects ING4 protein by western blot in over expressed cell lysates. This antibody was used at 1.0 µg/ml to detect ING4 expression in control (-) and transformed U2OS and HeLa cell lysates. A predominant band corresponding to p29 ING4 is only seen in lysates from transformed cells. Personnel Communication, Motoko Unoki, NCI, NIH. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          p29ING4 Antibody in Western Blot (WB)
          p29ING4 Antibody in Western Blot (WB)

          Product Details

          600-101-284

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:10,000
          -

          ELISA (ELISA)

          1:3,000-1:12,000
          -

          Immunoprecipitation (IP)

          1:500-1:3,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          This affinity purified antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 150-175 of Human p29 ING4 protein (Inhibitor of growth family, member 4). This sequence only shows homology to isoform 1 for ING4.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.28 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          0.02M potassium phosphate, pH 7.2, with 0.15M NaCl

          Contains

          0.01% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.

          Reactivity occurs against human p29 ING4 protein. The sequence is not present in isoforms 2 (see below). However, 100% homology is on record for this protein from human, dog, rat, and mouse. Cross reactivity with p29 ING4 protein from chimpanzee and chicken sources is also expected based on high homology (one amino acid change in sequence) by BLAST. Reactivity with p29 ING4 proteins from other sources is not known.

          Target Information

          The ING4 gene belongs to the ING (Inhibitor of Growth) family of tumor suppressors. All five members of the ING protein family identified to date contain a highly conserved plant homeodomain (PHD) finger motif at the C-terminal end, which is found in transcription factors that modulate chromatin structure. ING proteins have been shown to form transcriptional complexes leading to activation of responsive genes that mediate a wide range of cellular functions, including growth arrest, DNA repair, gene transcription, apoptosis, and senescence. The candidate tumor suppressor ING4 has a role in brain tumor pathogenesis, and is involved in regulating tumor growth and angiogenesis. Expression of ING4 is reduced in gliomas compared to normal brain tissue, and ING4 reduction correlates with progression from lower to higher grades of tumors. ING4 physically interacts with the p65 (RelA) subunit of NF-kB and represses transcription of NF-kB responsive genes. Inactivating mutations in ING4 transcripts have been described in various human cancer cell lines, and deletion of the ING4 locus has been detected in 10-20% of human breast cancer cell lines and tumors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: brain my036 protein; candidate tumor suppressor p33 ING1 homolog; Inhibitor of growth protein 4; p29ING4

          View more View less

          Gene Aliases: ING4; My036; p29ING4

          View more View less

          UniProt ID: (Human) Q9UNL4

          View more View less

          Entrez Gene ID: (Human) 51147

          View more View less

          Function(s)
          transcription coactivator activity protein binding zinc ion binding methylated histone binding
          Process(es)
          DNA replication protein acetylation apoptotic process DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator cell cycle arrest negative regulation of cell proliferation chromatin modification positive regulation of apoptotic process histone H3 acetylation histone H4-K5 acetylation histone H4-K8 acetylation histone H4-K12 acetylation histone H4-K16 acetylation negative regulation of transcription, DNA-templated negative regulation of growth
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0