Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Erythrocyte membrane protein band 4.2; Erythrocyte protein 4.2; erythrocyte protein band 4.2; erythrocyte surface protein band 4.2; P4.2; Protein 4.2
Gene Aliases: E42P; Epb4.2; EPB42; PA; SPH5
UniProt ID: (Human) P16452, (Mouse) P49222
Entrez Gene ID: (Human) 2038, (Mouse) 13828, (Rat) 362202
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support