Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a large, transmembrane receptor protein which may function in angiogenesis, lymphocyte homing, cell adhesion, or receptor scavenging. The protein contains 7 fasciclin, 16 epidermal growth factor (EGF)-like, and 2 laminin-type EGF-like domains as well as a C-type lectin-like hyaluronan-binding Link module. The protein is primarily expressed on sinusoidal endothelial cells of liver, spleen, and lymph node.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: common lymphatic endothelial and vascular endothelial receptor-1; fasciclin egf-like, laminin-type egf-like, and link domain-containing scavenger receptor-1; Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavenger receptor 1; FEEL-1; FEEL-1MS-1 antigen; MS-1 antigen; scavenger receptor FEEL-1; stabilin 1; Stabilin-1
Gene Aliases: CLEVER-1; FEEL-1; FEEL1; FELE-1; FEX1; KIAA0246; MFEEL-1; mKIAA0246; MS-1; SCARH2; STAB-1; STAB1
UniProt ID: (Human) Q9NY15
Entrez Gene ID: (Human) 23166, (Mouse) 192187, (Rat) 100363145
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support