Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • uNOS Antibodies

          AbboMax

          uNOS Polyclonal Antibody

          View all (3) uNOS antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite uNOS Polyclonal Antibody

          • Antibody Testing Data (1)
          uNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          uNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          uNOS Antibody (500-3934) in IHC (P)

          Immunohistochemistry: Human placenta tissue (FFPE) stained with Anti-uNOS antibody (Cat# 500-3934) at 1:1-200 for 10 min @ RT. Staining of formalin-fixed tissue requires boiling tissue sections in 10 mM Citrate Buffer, pH 6.0 for 10 min followed by cooling at RT for 20 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          uNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          500-3934

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-1 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          2-10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corrsponding to the internal segment of human uNos

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with proprietary stabilizer

          Contains

          0.01% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Wet ice

          Product Specific Information

          Positive control: Lung Tissue

          Cellular location: Cytoplasmic.

          Target Information

          NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities. NOS is classified under three types: neuronal NOS (nNOS) or brain NOS (bNOS); induciblr NOS (iNOS) or macrophage NOS (mNOS); and endothelial NOS (eNOS). This antibody reacts with iNOS, bNOS and eNOS.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bNOS; cNOS; Constitutive NOS; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; Endothelial NOS; HEP-NOS; Hepatocyte NOS; inducible nitric oxide synthase; Inducible NO synthase; Inducible NOS; MAC-NOS; Macrophage NOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxidase synthase; Nitric oxide synthase 1; nitric oxide synthase 1 (neuronal); nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); Nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; Nitric oxide synthase, brain; Nitric oxide synthase, endothelial; Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nNOS; NOS type I; NOS type II; NOS type III; NOS, type II; NOSIII; Peptidyl-cysteine S-nitrosylase NOS1; Peptidyl-cysteine S-nitrosylase NOS2

          View more View less

          Gene Aliases: 2310005C01Rik; 2310065A03Rik; Bnos; ECNOS; eNOS; HEP-NOS; i-NOS; IHPS1; INOS; Inosl; N-NOS; NC-NOS; nNOS; NO; NOS; Nos-1; Nos-2; Nos-3; NOS-I; NOS-II; NOS1; NOS2; NOS2A; NOS3

          View more View less

          UniProt ID: (Human) P29475, (Human) P35228, (Human) P29474, (Rat) P29476, (Mouse) Q9Z0J4, (Mouse) P29477, (Rat) Q06518, (Mouse) P70313, (Rat) Q62600

          View more View less

          Entrez Gene ID: (Human) 4842, (Human) 4843, (Human) 4846, (Rat) 24598, (Mouse) 18125, (Mouse) 18126, (Rat) 24599, (Mouse) 18127, (Rat) 24600

          View more View less

          Function(s)
          nitric-oxide synthase activity iron ion binding protein binding calmodulin binding FMN binding sodium channel regulator activity enzyme binding heme binding tetrahydrobiopterin binding arginine binding protein homodimerization activity ion channel binding cadmium ion binding flavin adenine dinucleotide binding NADP binding scaffold protein binding receptor binding metal ion binding actin monomer binding NADPH-hemoprotein reductase activity amino acid binding ATPase binding actin binding beta-catenin binding cAMP-dependent protein kinase regulator activity oxidoreductase activity protein kinase binding cadherin binding nitric-oxide synthase binding Hsp90 protein binding
          Process(es)
          angiogenesis ovulation from ovarian follicle in utero embryonic development blood vessel remodeling regulation of sodium ion transport regulation of the force of heart contraction by chemical signal regulation of systemic arterial blood pressure by endothelin arginine catabolic process nitric oxide biosynthetic process mitochondrion organization nitric oxide mediated signal transduction regulation of blood pressure negative regulation of cell proliferation response to heat negative regulation of platelet activation negative regulation of muscle hyperplasia smooth muscle hyperplasia removal of superoxide radicals lung development positive regulation of guanylate cyclase activity lipopolysaccharide-mediated signaling pathway response to fluid shear stress negative regulation of potassium ion transport endothelial cell migration cell redox homeostasis positive regulation of angiogenesis negative regulation of blood pressure positive regulation of vasodilation regulation of blood vessel size regulation of nitric-oxide synthase activity negative regulation of hydrolase activity negative regulation of calcium ion transport oxidation-reduction process negative regulation of extrinsic apoptotic signaling pathway via death domain receptors response to hypoxia positive regulation of leukocyte mediated cytotoxicity innate immune response in mucosa superoxide metabolic process circadian rhythm response to bacterium negative regulation of gene expression peptidyl-cysteine S-nitrosylation prostaglandin secretion cellular response to drug regulation of cell proliferation negative regulation of protein catabolic process defense response to bacterium regulation of cellular respiration regulation of insulin secretion defense response to Gram-negative bacterium positive regulation of killing of cells of other organism cellular response to lipopolysaccharide cellular response to interferon-gamma interleukin-6 secretion interleukin-8 secretion regulation of cytokine production involved in inflammatory response inflammatory response response to lipopolysaccharide regulation of protein kinase activity signal transduction response to estradiol positive regulation of apoptotic process negative regulation of smooth muscle cell proliferation cellular response to mechanical stimulus cellular response to transforming growth factor beta stimulus muscle contraction striated muscle contraction female pregnancy aging regulation of heart contraction response to organonitrogen compound response to lead ion response to organic cyclic compound response to activity response to nutrient levels negative regulation of peptidyl-serine phosphorylation response to vitamin E multicellular organismal response to stress positive regulation of histone acetylation response to nicotine exogenous drug catabolic process negative regulation of apoptotic process response to peptide hormone response to estrogen response to ethanol negative regulation of heart contraction positive regulation of transcription, DNA-templated negative regulation of vasoconstriction positive regulation of transcription from RNA polymerase II promoter negative regulation of insulin secretion behavioral response to cocaine negative regulation of cytosolic calcium ion concentration negative regulation of serotonin uptake regulation of sensory perception of pain cellular response to growth factor stimulus response to nitric oxide cellular response to epinephrine stimulus positive regulation of the force of heart contraction positive regulation of long-term synaptic potentiation positive regulation of adrenergic receptor signaling pathway involved in heart process positive regulation of neuron death positive regulation of sodium ion transmembrane transport myoblast fusion negative regulation of calcium ion transport into cytosol neurotransmitter biosynthetic process regulation of neurogenesis regulation of cardiac muscle contraction regulation of ryanodine-sensitive calcium-release channel activity negative regulation of adrenergic receptor signaling pathway involved in heart process regulation of calcium ion transmembrane transport via high voltage-gated calcium channel regulation of cardiac conduction response to mechanical stimulus response to fructose response to metal ion response to cytokine cellular response to heat response to statin response to drug response to amino acid response to axon injury response to corticosterone response to hyperoxia response to lipoprotein particle retina development in camera-type eye bone development cellular response to magnesium ion cellular response to cAMP cellular response to tumor necrosis factor cellular response to organic cyclic compound response to candesartan response to peptide response to ketone response to salt endothelial cell proliferation G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger response to hormone glycoside metabolic process cGMP-mediated signaling response to tumor necrosis factor intracellular signal transduction response to immobilization stress wound healing retinoic acid receptor signaling pathway cellular response to lithium ion cellular response to vitamin D cellular response to interleukin-1 cellular response to redox state response to dexamethasone
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0