404 Not Found
Not Found
The requested URL /msheaderfooter/tf/header/anonymous/en/br/.shtml was not found on this server.
Profile more biomarkers with less sample
Invitrogen ProcartaPlex immunoassays are antibody-based, magnetic bead reagent kits and panels for multiplex protein quantitation using the Luminex instrument platform. The assays cover more than 600 cytokines, chemokines, growth factors, and other protein targets from human, mouse, rat, non-human primate, canine, and other species. Select from 90+ preconfigured multiplex panels (2- to 80-plex), or blend singleplex kits to create custom multiplex assays.
ProcartaPlex multiplex assay options
About ProcartaPlex assays
ProcartaPlex multiplex immunoassay kits
ProcartaPlex multiplex immunoassays make use of Luminex xMAP (multi-analyte profiling) technology to enable the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 µL sample of plasma, serum, cell culture supernatants, and other bodily fluids. Luminex technology uses differentially dyed capture beads for each target in a multiplex ELISA like assay. ProcartaPlex immunoassays enable the simultaneous analysis of multiple proteins in a single sample from a broad range of biological sources. They combine the efficiencies of multiplexing with the accuracy, sensitivity, reproducibility, and simplicity of ELISA.
Key features of ProcartaPlex immunoassays:
  • Multi-analyte detection with a large dynamic range
  • All ProcartaPlex assays are individually tested for specificity and detection of native endogenous protein
  • ProcartaPlex Preconfigured panels are extensively tested for combinability, interference, and cross-reactivity
  • Scalable and reproducible performance regardless of plex size
  • Magnetic bead format for reliable washing
  • Largest panel for quantitation of up to 80 analyte proteins
  • New 384-well format with lyophilized beads options
ProcartaPlex workflow
ProcartaPlex multiplex assay kits are performed in much the same way as ELISAs. The steps are similar, with the exception that antibody-coated beads are used to capture the analyte, instead of having capture antibodies attached directly to the plate. Each kit provides standards of known concentration so that a standard curve can be established. After incubation on a shaker, the beads are washed by putting the 96-well plate on a flat magnet for 30 seconds, after which the fluid is discarded by flicking the wells or by using an automated plate washer. The magnet is removed, and the beads are resuspended in the detection antibody. Another incubation and wash are followed by the addition of streptavidin–R-phycoerythrin (SAPE). The beads are then washed and are ready to analyze. Access streamlined analysis with the ProcartaPlex Analysis App.
ProcartaPlex multiplex immunoassays for the Luminex platform are performed in a similar way as traditional ELISAs but with the ability to quantitate up to 80 targets from a single sample.
General workflow for the Invitrogen ProcartaPlex multiplex immunoassay
Over 600 analytes are available for human, mouse, rat, non-human primate (NHP), canine, and porcine samples. Below are analytes that are available either in preconfigured multiplex panels or available as Simplex Kits for building your own assay. Note that analytes in the tables below marked with * are only available as part of a panel.
Design your own assays
ProcartaPlex assays offer the ultimate flexibility; you can select your own Simplex assays and mix them yourself, or design the panel of your choice with our online Invitrogen ProcartaPlex Panel Configurator. Select your analytes of interest, and immediately receive recommendations for the most effective solutions for your panel needs. Each customized panel is formulated and supplied with all the necessary reagents to perform the assay, including premixed capture beads, premixed detection reagents, protein standards, and buffers. If you are not sure which solution is right for you, use the panel configurator to discover custom and catalog options for your targets of interest.
Use the ProcartaPlex Panel Configurator and Selection Tool to select analytes of interest and build your panel.
Inflammation and cytokine storm assays
Measure biomarkers of cytokine storm and inflammation.
SARS-CoV-2 Serology Assays
Rapidly detect antibodies (immunoglobulins, IgG and IgM) in serum against the SARS-CoV-2 coronavirus.
Featured products
Matched gene expression and protein panels
Gain deeper insight with complementary and highly correlative multiplex panels. You can measure gene expression and protein on a single platform and have confidence in your results.
Understand the effects of cytotoxins in biological systems
Analyze 11 biomarkers (including FDA recommended markers) to monitor kidney disease progression, drug induced kidney toxicity, and nephrotoxicity.
Access the largest multiplex panel on the market
You can expand your search and analyze up to 80 protein targets in a single well. You only need 25 µL of plasma or serum sample for this assay.
QuantiGene Plex gene expression assays
Gain deep insight with complementary and highly correlative gene expression panels.
Invitrogen ELISA Kits
Quantitate single analytes with highly verified Invitrogen ELISA kits.
Immunoassay research areas
Explore our immunoassays for different research areas, including immune-oncology, neurobiology, isotyping and immunoglobulin characterization, and more.
ProQuantum High Sensitivity Immunoassays
Detect very low levels of proteins using as little as 2–5 μL of sample.
For Research Use Only. Not for use in diagnostic procedures.
404 Not Found
Not Found
The requested URL /msheaderfooter/tf/footer/anonymous/en/br/.tagManagerFooterOff.shtml was not found on this server.