The requested URL /msheaderfooter/tf/header/anonymous/en/br/.shtml was not found on this server.
Thermo Scientific offers a range of storage tube accessories to enhance the security and organization of your samples when using our tubes. Our sample storage tube accessories are designed to work with our tubes in support of your lab’s workflow.
Whether to protect an individual cryotube with Cryoflex or to protect a cryotube loaded CryoCane with CryoSleeve, our storage tube protection accessories safeguard your samples in liquid or vapor phase nitrogen during cryopreservation and shipping.
Our portfolio of sample storage tubes includes various capping options and can be ordered with the capping system that best suits your needs. Additionally, we offer a range of caps and seals that can be purchased separately to support your specific needs. Matrix and Nunc caps are available in a range of formats and colors to allow for your manual or automated storage needs.
Cryogenic storage is a harsh environment that can cause writing and label to deteriorate and become unreadable over time. It's important to use cryogenically compatible markers, labels, and pens to ensure you will always be able to track and locate your precious samples when you need them. Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific Cryo Marker Pen Set and 25 x 50mm cloth labels specifically designed for use at ultra-low temperatures without shrinking or peeling.
This product is intended for general laboratory use unless otherwise specified. It is the customer’s responsibility to ensure that the performance of the product is suitable for the specific use or application.