The Thermo Scientific OptaMax NG ion source easily analyzes small molecules by enabling improved positional adjustment and optimized flow-rate dependent source settings. The exhaust design captures spray more efficiently, leading to less peak tailing and carryover. The exhaust pipe mates automatically with source housing.