Search Thermo Fisher Scientific
Water is a precious resource and its scarcity has already influenced the quality of our lives. Water regulations are critical to enforce the preservation of our environment and water resources so we have safe water to drink and clean water to use as the supply for drinking water and irrigation. On this page, we provide solutions to comply with water regulations set in the U.S. and Europe.
The U.S. Safe Drinking Water Act (SDWA) regulates the drinking water and requires the U. S. EPA to set the National Primary Drinking Water Regulations (NPDWR) and National Secondary Drinking Water Regulations (PSDWR). The U.S. Clean Water Act (CWA) regulates the discharge of pollutants into U.S. surface waters and sets quality standards for surface waters. Under the CWA, the U.S. EPA has implemented pollution control programs that include wastewater standards for industry and water quality standards for all contaminants in surface waters. To protect water resources and guarantee a safe supply of drinking water, both drinking water standards and effluent (wastewater) discharge standards must be enforced. Variety of approved analytical methods are required to used for analysis of contaminants in water, such as the EPA methods, ASTM methods, Standard Methods, and others. These methods have been continually been updated with newer technologies and higher sensitivities to meet the needs of new and stricter regulatory standards. Meanwhile, Alternate Test Procedures (ATP) have been approved by the EPA for contaminant analysis. Stay current with alternative technologies and methodologies approved for compliance monitoring through the EPA’s Alternative Test Procedures (ATP) program.
The EU Water Framework Directive (WFD) was introduced in August 2013 amending the EU directives 2000/60/EC and 2008/105/EC. It is a directive which:
Although the directive acts at the whole EU level, member states can choose to apply an environmental quality standard (EQS) for an alternative matrix or, where relevant, an alternative biota taxon. In addition, the new directive encourages the development of novel monitoring methods such as passive sampling and other tools.
CIP 2 UK regulations investigates the occurrence, sources and removal of trace substances in waste water treatment facilitiy effluent. This regulation helps to establish priorities for remediative action to ensure surface waters meet new Environmental Quality Standards (EQS).
The primary objective of CIP 2 is to identify and characterize sites where EQS levels are breached. In the program 70 priority substances were determined from 162 sewage treatment works (STW) effluents. 11 pharmaceuticals were also identified as priority monitoring candidates. It is important to note that EQS is defined for only 3 pharmaceutical compounds Diclofenac, E2 and EE2. All substances selected for monitoring analysis were detected previously in waste water effluent samples. The determined environmental concentrations of many priority substances in effluent exceeded EQS.