The requested URL /msheaderfooter/tf/header/anonymous/en/us/.shtml was not found on this server.
Powered by proven technology, the Applied Biosystems QuantStudio 7 Pro Dx Real-Time PCR System is a standalone qPCR platform that is versatile, highly expandable, and easy to use. Designed to support in vitro diagnostic (IVD) applications, the QuantStudio 7 Pro Dx system incorporates smart features such as hand-free functionality. Whether your laboratory is running routine assays or developing new ones, the QuantStudio 7 Pro Dx can support your work.
With sensitivity down to 1.5-fold in singleplex reactions and 10 logarithmic units of linear dynamic range, the QuantStudio 7 Pro Dx system supports the development of molecular diagnostic (MDx) assays. The compact footprint, software features, and up to six-color multiplexing helps enable laboratory productivity and innovation.
The automated QuantStudio 7 Pro Dx Real-Time PCR System is an in vitro diagnostic device intended to perform fluorescence-based PCR to provide detection of nucleic acid sequences in human-derived specimens.
The QuantStudio 7 Pro Dx Real-Time PCR System is intended to be used by trained professionals in combination with in vitro diagnostic assays.
Key features of the QuantStudio 7 Pro Dx system include:
Enhanced security for confidence
Security, auditing, and e-signature (SAE) functionalities assist in compliance with regulatory requirements and support multiple clients. SAE settings that can be applied to multiple instruments on the same network, allow better control for your IT department.
Easy-to-use, powerful software
The included software is designed to guide users through workflow steps for test development* or in vitro diagnostics. Suitable for clinics, hospitals, and diagnostic test developers, the QuantStudio 7 Pro Dx software enables users to set up a run, lay out assays, control the instrument, and conduct plate analysis on an easy-to-use touchscreen interface.
Assay-driven qPCR workflows
For ease of use and error reduction, create and manage assay definitions files (ADFs) for a specific assay, panel, or test. The ADFs incorporate all of the qPCR setup and analysis parameters required for a particular assay. Once installed on the instrument, operators select a specific ADF to run, helping reduce setup time on the device and user error. ADFs help reduce risk of unauthorized use and accidental or intentional misuse by embedding regulatory status of an assay in the file.
Compact, stand-alone instrument
Featuring a small footprint without the requirement for an attached computer, the graphical user interface (GUI) enables end-to end workflows from the touchscreen. The SAE administrative console can be run in server configuration to free up valuable bench space.
Green features enhance efficiency
Use up to 54% less material, generate 91% less waste during calibration, and use up to 26% less energy than comparable instruments.
6-color multiplexing
A six-color, decoupled OptiFlex filter system with bright white LED is engineer-calibrated during installation to enable optimal performance.
Smart features
Hands-free functionality includes the ability to perform basic operations using voice commands. The QuantStudio 7 Pro Dx system also includes smart support and smart help to report issues or request instrument services to reduce downtime.
Interchangeable Block Options
Achieve maximum flexibility with interchangeable block options of 96-well 0.2 mL and 384-well. With interchangeable blocks, test development and in vitro diagnostics are possible on the same instrument. Increasing test volumes can be accommodated if protocol is validated on both 96-and 384-well blocks.
Instrument verification or calibration plate |
Cat. No. |
---|---|
RNase P 96-Well Instrument Verification Plate |
|
96‑Well Region of Interest (ROI) and Background Plates (2 plates) |
|
96‑Well 0.2‑mL 10‑Dye Spectral Calibration Kit (contains all 3 spectral calibration plates listed below) |
|
96‑Well 0.2‑mL Spectral Calibration Plate 1 (FAM, VIC, ROX, and SYBR dyes) |
|
96‑Well 0.2‑mL Spectral Calibration Plate 2 (ABY, JUN, and MUSTANG PURPLE dyes) |
|
96‑Well 0.2‑mL Spectral Calibration Plate 3 (TAMRA, NED, and Cy5 dyes) |
|
RNase P 384-Well Instrument Verification Plate | 4432500 |
384-WellRegion of Interest (ROI) and Background Plates | 4432482 |
384-Well 10-Dye Spectral Calibration Kit (contains the 2 spectral calibration plates listed below) |
A52523 |
384-Well Spectral Calibration Plate 1 (FAMTM, VICTM, ROXTM, SYBRTM, and TAMRATM dyes) | A52439 |
384-Well Spectral Calibration Plate 2 (ABYTM, JUNTM, MUSTANG PURPLETM, NEDTM, and CyTM 5 dyes) |
A52440 |
Technical specifications |
|
---|---|
Reaction volume |
96-well, 0.2-mL block: 10-100 μL, 384-well |
Excitation source |
Bright-white LED |
Optical detection |
6 decoupled filters, complementary metal oxide semiconductor (CMOS) |
Excitation/detection range |
450-680 nm/500-730 nm |
Temperature range |
4-99.9°C |
Maximum block ramp rate |
6.5°C/sec |
Average sample ramp rate |
3.66°C/sec |
Temperature uniformity |
0.5°C |
Temperature accuracy |
0.25°C |
Heating and cooling method |
Peltier |
Independent temperature zones |
6 VeriFlex zones (5°C zone to zone) |
Chemistries |
Both fast and standard |
Run times |
<30 min (fast mode) |
System dyes |
Applied Biosystems FAM, SYBR, VIC, ABY, NED, TAMRA, JUN, ROX, Mustang Purple, and Cy5 dyes |
Multiplex |
5-plex with 1 passive reference; 6-plex with no passive reference |
Sensitivity |
1 copy; detect differences as small as 1.5-fold in target quantities in singleplex |
Dynamic range |
10 orders of magnitude of linear dynamic range |
Touchscreen |
12-inch capacitive touchscreen with real-time application viewing |
Online ecosystem |
Free, Connect cloud-based platform with cloud-enabled systems |
Security and auditing features |
- Integrated tools to assist with compliance - The audit function is always enforced |
Footprint (W x D x H) |
53 cm x 55 cm x 34 cm |
Weight |
38 kg |
For In Vitro Diagnostic Use.
This instrument may not be available in your region.