1. Pre-incubate test cells in appropriate microplate wells
Cell membrane integrity—based viability assay
The CyQUANT Direct assays are based on a cell-permeant DNA-binding dye in combination with a background suppression reagent. The DNA-binding dyes stains all cells, but in cells with compromised cell membranes the fluorescence is masked by the background suppressors. Thus, the fluorescent signal in the CyQUANT Direct assays are proportional to the number of live cells. Live cells can also be counted using an imaging system. The assays are available in either green or red emission versions.
This protocol can be used for:
- Quantifying live cells using a microplate reader
This protocol should not be used for:
- Flow cytometry
You will need the following for this protocol:
Assay protocol
2. Add an equal volume of CyQUANT Direct 2X detection reagent to microplate wells (see instructions for making 2X detection reagent and recommended well volumes)
3. Incubate for 60 minutes at 37°C
![]() | 4. Read fluorescence using excitation/emission |
5. Plot a standard curve of fluorescence vs. cell number to generate quantitative results
Protocol tips
- The assay can tolerate a variety of cell culture media components including phenol red and up to 10% serum
- Plot a standard curve of fluorescence vs. cell number to generate quantitative results
- When prepared aseptically, the 2X detection reagent is generally stable for up to 24 hours at room temperature
1. Combine the following in a 15 mL tube:
- Hank’s buffered saline solution, PBS, or cell culture medium—11.7 mL
- CyQUANT Direct nucleic acid stain—48 µL
- CyQUANT Direct background suppressor— 240 µL
2. Mix well and let stand at room temperature until ready for use. See table below for example volumes; CyQUANT Direct signal is stable for 7 hours
Format | Volume of cells + medium/well | Volume of 2X detection reagent/well |
---|---|---|
96-well plate | 100 μL | 100 μL |
384-well plate | 40 μL | 40 μL |
1,536-well plate | 5 μL | 2 μL |