Reverse Osmosis Membranes
Reverse Osmosis Membranes
Actual product may vary
Thermo Scientific™

Reverse Osmosis Membranes

Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
09.2006Reverse Osmosis membrane with integrated pretreatment, 6L/hr
22.0087Reverse Osmosis membrane
09.2003Reverse Osmosis membrane with integrated pretreatment, 3L/hr
22.0046Reverse Osmosis membrane, for use with LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20
Catalog number 09.2006
Price (USD)
583.00
Each
Add to cart
Description:
Reverse Osmosis membrane with integrated pretreatment, 6L/hr
Price (USD)
583.00
Each
Add to cart
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.

Use Membranes with the Following Barnstead Systems:

09.2003:

  • Reverse osmosis membrane with integrated pretreatment, 3L/hr for Smart2Pure™ systems

09.2006:

  • Reverse osmosis membrane with integrated pretreatment, 6L/hr for Smart2Pure systems

22.0046:

  • LabTower™ EDI 15 requires two of these RO membranes
  • LabTower™ RO 20 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • LabTower™ TII 20 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • Pacific™ RO 3, 7, 12 require one of these RO membranes
  • Pacific RO 20 requires two of these RO membranes
  • Pacific™ TII 3, 7, 12 require one of these RO membranes
  • Pacific TII 20 requires two of these RO membranes

22.0087:

  • LabTower™ EDI 30 requires two of these RO membranes
  • LabTower™ RO 40 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • LabTower TII 40 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • Pacific RO 40 requires two of these RO membranes
  • Pacific TII 40 requires two of these RO membranes
Specifications
DescriptionReverse Osmosis membrane with integrated pretreatment, 6L/hr
For Use With (Equipment)Smart2Pure
TypeReverse Osmosis Membrane (pretreated)
Unit SizeEach