Search Thermo Fisher Scientific
Catalog Number | Description | Volume (Metric) | Tip Style | Fit for Purpose | Unit Size |
---|---|---|---|---|---|
2149P-05-RTPK also known as 2149P-05-RT | ART Barrier, Filtered, Sterile, Rack | 20 μL | Low Retention | Sterile & PCR Ready | Pack of 960 |
2139-RT | ART Barrier, Filtered, Sterile, Rack | 10 μL | Standard | Sterile & PCR Ready | Case of 4800 |
2139-RTPK also known as 2139-RT | ART Barrier, Filtered, Sterile, Rack | 10 μL | Standard | Sterile & PCR Ready | Pack of 960 |
2139-05-RT | ART Barrier, Filtered, Sterile, Rack | 10 μL | Low Retention | Sterile & PCR Ready | Case of 4800 |
2139-05-RTPK | ART Barrier, Filtered, Sterile, Rack | 10 μL | Low Retention | Sterile & PCR Ready | Pack of 960 |
2149P-RT | ART Barrier, Filtered, Sterile, Rack | 20 μL | Standard | Sterile & PCR Ready | Case of 4800 |
2149P-RTPK also known as 2149P-RT | ART Barrier, Filtered, Sterile, Rack | 20 μL | Standard | Sterile & PCR Ready | Pack of 960 |
2149P-05-RT | ART Barrier, Filtered, Sterile, Rack | 20 μL | Low Retention | Sterile & PCR Ready | Case of 4800 |
2069-RT | ART Barrier, Filtered, Sterile, Rack | 200 μL | Standard | Sterile & PCR Ready | Case of 4800 |
2069-RTPK also known as 2069-RT | ART Barrier, Filtered, Sterile, Rack | 200 μL | Standard | Sterile & PCR Ready | Pack of 960 |
2069-05-RT | ART Barrier, Filtered, Sterile, Rack | 200 μL | Low Retention | Sterile & PCR Ready | Case of 4800 |
2069-05-RTPK also known as 2069-05-RT | ART Barrier, Filtered, Sterile, Rack | 200 μL | Low Retention | Sterile & PCR Ready | Pack of 960 |
2070-RT | ART Barrier, Filtered, Sterile, Rack | 300 μL | Standard | Sterile & PCR Ready | Case of 4800 |
2070-RTPK also known as 2070-RT | ART Barrier, Filtered, Sterile, Rack | 300 μL | Standard | Sterile & PCR Ready | Pack of 960 |
2070-05-RT | ART Barrier, Filtered, Sterile, Rack | 300 μL | Low Retention | Sterile & PCR Ready | Case of 4800 |
2070-05-RTPK also known as 2070-05-RT | ART Barrier, Filtered, Sterile, Rack | 300 μL | Low Retention | Sterile & PCR Ready | Pack of 960 |
Perform applications accurately and consistently — without compromising protection — with the Thermo Scientific™ ART™ Barrier Reload System, which provides an environmentally responsible solution without compromising sterility. With offset wells that allow the tips to stack, this advanced system reduces waste, and saves benchtop and storage space. The ART portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in your laboratory.