SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene)
SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene)
Thermo Scientific™

SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene)

Thermo Scientific Pierce SMPT is a long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form strongRead more
Have Questions?
Catalog number 21558
Price (USD)
358.00
Each
Add to cart
Request bulk or custom format
Price (USD)
358.00
Each
Add to cart
Thermo Scientific Pierce SMPT is a long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form strong (hindered) yet cleavable (reducible) disulfide bonds with cysteine sulfhydryls.

Features of SMPT:

Reactive groups: NHS ester and pyridyldithiol
Reactive towards: amino and sulfhydryl groups
• Cleavable with disulfide reducing agents
• Water-insoluble (dissolve first in DMF or DMSO)
• Contains a hindered disulfide bond; has formed immunotoxins with improved stability
• In vitro, an SMPT conjugate was as effective as conjugates formed with SPDP and 2-IT
• Compare to other varieties of SPDP-type reagents, including pegylated forms

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
SPDP (succinimidyl 3-(2-pyridyldithio)propionate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine-Sulfhydryl
CleavableBy Thiols
DescriptionSMPT
Molecular Weight (g/mol)388.46
PEGylatedNo
Spacer Arm Length11.2 Å
Cell PermeabilityYes
Shipping ConditionAmbient
Product LinePierce™
Labeling MethodChemical Labeling
Crosslinker TypeHeterobifunctional
Reactive MoietyNHS Ester, Pyridyldisulfide
SpacerMedium (10 to 30 Å)
FormPowder
Quantity50 mg
SolubilityDMF, DMSO
FormatStandard
Water SolubleNo
Product TypeCrosslinker
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at 4°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene)?

SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.