Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Actual product may vary
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Thermo Scientific™

Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles

Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Chemical LabelUnit Size
2436-0502PK
also known as 2436-0502
500 mLEthyl AlcoholPack of 4
2436-0503500 mLMethanolCase of 24
2436-0503PK
also known as 2436-0503
500 mLMethanolPack of 4
2436-0504500 mLIsopropanolCase of 24
2436-0504PK
also known as 2436-0504
500 mLIsopropanolPack of 4
2436-0505500 mLDistilled WaterCase of 24
2436-0505PK
also known as 2436-0505
500 mLDistilled WaterPack of 4
2436-0502500 mLEthyl AlcoholCase of 24
2436-0506500 mLSodium HypochloriteCase of 24
2436-0506PK
also known as 2436-0506
500 mLSodium HypochloritePack of 4
2436-0501500 mLAcetoneCase of 24
2436-0501PK
also known as 2436-0501
500 mLAcetonePack of 4
2436-10051000 mLDistilled WaterCase of 12
2436-1005PK
also known as 2436-1005
1000 mLDistilled WaterPack of 2
2436-10011000 mLAcetoneCase of 12
2436-1001PK
also known as 2436-1001
1000 mLAcetonePack of 2
Catalog number 2436-0502PK
also known as 2436-0502
Price (USD)
65.00
Pack of 4
Add to cart
Capacity (Metric):
500 mL
Chemical Label:
Ethyl Alcohol
Unit Size:
Pack of 4
Price (USD)
65.00
Pack of 4
Add to cart

Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady, no sputter stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.

  • Bottle and tubulation molded in one piece to prevent chemical splash when removing closure and contamination when filling
  • Unique 38mm colored vented closure designed to prevent dripping and self-dispensing
  • Sturdy polypropylene closures (HDPE on Sodium Hypochloride bottles) with integrated vent, color coded for easy identification
  • Color-coded bar corresponds to closure, preventing closure mix-ups and cross-contamination
  • Prelabeled with ICS name and formula, as well as NFPA symbols for chemical handling, hazard, care and storage of six commonly-used chemicals
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Tip can be cut back to provide increased flow
.

Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
 
Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
DescriptionRTK Wash Bottle
MaterialLDPE
No. per Case24
No. per Pack4
Closure ColorWhite
UnitaryYes
Certifications/ComplianceMeet OSHA requirements for workplace chemical container labeling.
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeWash Bottle
Chemical LabelEthyl Alcohol
Closure MaterialPolypropylene
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizePack of 4