Thermo Scientific Pierce Sulfo-SHPP, sulfosuccinimidyl-3 -(4-hydroxypheynyl) propionate, also known as water-soluble Bolton-Hunter Reagent, conjugates tyrosine-like functional groups to primary aminesRead more
Have Questions?
Catalog number 27712
Price (USD)
644.00
Each
Add to cart
Request bulk or custom format
Price (USD)
644.00
Each
Add to cart
Thermo Scientific Pierce Sulfo-SHPP, sulfosuccinimidyl-3 -(4-hydroxypheynyl) propionate, also known as water-soluble Bolton-Hunter Reagent, conjugates tyrosine-like functional groups to primary amines to increase the number of tyrosyl groups that can be iodinated by iodine-125 labeling procedures.
Features of Sulfo-SHPP:
• Iodinate before or after coupling Sulfo-SHPP to the molecule of interest • Useful for introducing tyrosyl groups on proteins sensitive to small amounts of organic solvent • Useful for labelling cell surfaces without exposing the cells to membrane permeable solvents
For Research Use Only. Not for use in diagnostic procedures.
SolubilityDMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or DyeIodine-125
SpacerShort
Label TypeRadioisotope
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C. Product is shipped at ambient temperature.
Frequently asked questions (FAQs)
Can you provide the shelf-life for Sulfo-SHPP (Water-Soluble Bolton-Hunter Reagent)?
Sulfo-SHPP (Water-Soluble Bolton-Hunter Reagent) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.