HyperSep™ PFAS Cartridges
HyperSep™ PFAS Cartridges
Actual product may vary
HyperSep™ PFAS Cartridges
HyperSep™ PFAS Cartridges
HyperSep™ PFAS Cartridges
HyperSep™ PFAS Cartridges
Thermo Scientific™

HyperSep™ PFAS Cartridges

HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightDescriptionFor Use With (Application)Particle Size
60107-500-6-HLD500 mgHyperSep PFAS SPE, 500 mg, 6 mL, HLDEPA method 537, extraction of PFAS from water20 to 40 μm
60107-150-6-WAX150 mgHyperSep PFAS SPE, 150 mg, 6 mL, WAXEPA method 1633, extraction of PFAS from water, tissue, soil, or biosolids55 to 65 μm
60107-20050-WAXGCB250 mg200 mg Polymeric WAX on top of 50 mg of graphitized carbon black (GCB)EPA method 1633, extraction of PFAS from water55 to 65 μm and 120/400 mesh
60107-50200-GCBWAX250 mgHyperSep PFAS SPE, Dual PHASE, 50 mg GCB, 200 MG WAX, 6 mLEPA method 1633, extraction of PFAS from tissue, soil, or biosolids55 to 65 μm and 120/400 mesh
60107-500-6-WAX500 mgHyperSep PFAS SPE, 500 mg, 6 mL, WAXEU and other regulations, extraction of PFAS from water55 to 65 μm
60107-30-3-WAX30 mgHyperSep PFAS SPE, 30 mg, 3 mL, WAXEU and other regulations, extraction of PFAS from water55 to 65 μm
60107-200-6-WAX200 mgHyperSep PFAS SPE, 200 mg, 6 mL, WAXEPA method 1633 (alternative), extraction of PFAS from water, tissue, soil, or biosolids55 to 65 μm
GCB-1010 gHyperSep LOOSE CARBONEPA method 1633, extraction of PFAS from water, tissue, soil, or biosolids in conjunction with SPE cartridge55 to 65 μm and 120/400 mesh
Catalog number 60107-500-6-HLD
Price (USD)
260.00
Pack of 30
Add to cart
Bed Weight:
500 mg
Description:
HyperSep PFAS SPE, 500 mg, 6 mL, HLD
For Use With (Application):
EPA method 537, extraction of PFAS from water
Particle Size:
20 to 40 μm
Price (USD)
260.00
Pack of 30
Add to cart
HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination. The range of selectivity’s and formats are designed to meet the requirements for main environmental testing methods by extracting and concentrating samples from drinking water, ground water, surface water, wastewater, soil, food and other complex matrices.

Per- and polyfluorinated alkyl substances (PFAS) are among the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances is rapidly growing. SPE solutions are a fundamental part of the analytical workflow for analysis of PFAS compounds.

Thermo Scientific provides a range of products to address PFAS testing requirements e.g. EPA methods: 1633, 537.1, 533.

Specifications
DescriptionHyperSep PFAS SPE, 500 mg, 6 mL, HLD
Bed Weight500 mg
For Use With (Application)EPA method 537, extraction of PFAS from water
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size20 to 40 μm
Product LineHyperSep
SorbentPolymeric DVB
TypeSolid Phase Extraction Column
Volume (Metric)6 mL
Quantity30/Pk.
Unit SizePack of 30