GlycoLink™ IP Kit
GlycoLink™ IP Kit
Thermo Scientific™

GlycoLink™ IP Kit

The Thermo Scientific GlycoLink IP Kit provides components for effective immunoprecipitation (IP and co-IP) based on small-scale, covalent, affinity-resin immobilizationRead more
Have Questions?
Catalog number 88943
Price (USD)
724.00
Each
Add to cart
Request bulk or custom format
Price (USD)
724.00
Each
Add to cart
The Thermo Scientific GlycoLink IP Kit provides components for effective immunoprecipitation (IP and co-IP) based on small-scale, covalent, affinity-resin immobilization of antibodies and other glycoproteins via oxidized sugar groups.

Features of the GlycoLink IP Kit:

Efficient immobilization—couple 2 to 10 μg of oxidized antibody or other glycoprotein to 20 μL of resin in 2 hours or less
Stable immobilization—resonance structure of the hydrazone bonds are sufficiently stable so the antibody remains coupled during elution
Preserves binding function—immobilizes IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target
Versatile applications—immobilize any molecule that contains oxidizable sugars, including glycoproteins for use in purifying protein interaction binding partners

The GlycoLink Kit uses hydrazide-activated UltraLink Resin to immobilize and perform 25 IP or Co-IP assays with different antibodies or glycoprotein ligands. The method is ideal for polyclonal antibodies, which typically have abundant carbohydrates (glycosylation) on their Fc portions. Monoclonal antibodies that contain adequate carbohydrates are also effective. Antibodies immobilized by this hydrazide-to-carbohydrate method have unobstructed antigen-binding sites and optimal purification capability. The prepared glycoprotein or antibody columns can be regenerated and reused at least five times for immunoprecipitation, co-immunoprecipitation or pull-down affinity-purification without significant loss in binding capacity.

Applications:
• Immunoprecipitation followed by electrophoresis and excision of the protein for mass spectrometry or sequencing
• Immunoprecipitation and SDS-PAGE analysis of a protein with a molecular weight similar to the antibody's heavy or light chain
• Identification of binding partners of immobilized glycoprotein
• Co-immunoprecipitation for determining protein-protein interactions

Includes:
Kit contains resin, buffers and reagents for immobilization; desalting columns; IP lysis/wash and elution buffers for immunoprecipitation; microcentrifuge spin columns and collection tubes; and loading buffer for SDS-PAGE

The GlycoLink IP Kit method results in covalent attachment of oxidized sugar groups of an IP antibody (or any other glycoprotein with oxidizable carbohydrate groups) to the hydrazide resin. The antibody resin is then incubated with the sample that contains the protein antigen of interest, allowing the antibody-antigen complex to form. After washing to remove non-bound components of the sample, the antigen is recovered by dissociation from the antibody with elution buffer supplied in the kit. The entire procedure is performed in a microcentrifuge spin cup, allowing solutions to be fully separated from the resin with a brief centrifugation. Only the antigen is eluted by the procedure, enabling it to be identified and further analyzed without interference from antibody fragments.

More Product Data
Improved immobilization and conjugation of glycoproteins
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)2 to 10 μg Oxidized Antibody or Glycoprotein Coupling to 20 μL Resin
AssayAffinity Purification
For Use With (Equipment)Microcentrifuge
Product LineGlycoLink™
Quantity1 Kit
For Use With (Application)Immunoprecipitation (IP)
TargetNot Target-Specific
TypeImmunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For25 Reactions
FormatKit
Unit SizeEach
Contents & Storage
Sufficient For: 25 IP/Co-IP reactions, each using 20 μL antibody-coupled resin
• UltraLink Hydrazide Resin, 0.5 mL
• GlycoLink Coupling Buffer, 50 mL
• Aniline, 0.2 mL
• Sodium meta-periodate, 0.5 g
• Wash Buffer, 60 mL
• Zeba Spin Desalting Columns,0.5 mL, 25 columns
• IP Lysis/Wash Buffer, 50 mL
• PBS Pack (makes 500 mL), 1 pack
• Elution Buffer, pH 2.8, 50 mL
• Lane Marker Sample Buffer, Non-reducing, (5X), 5 mL
• Pierce Spin Columns-Screw Caps, 25 columns
• Collection Tubes, 2 mL, 100 tubes

Store at 4°C.

Frequently asked questions (FAQs)

What is the difference between GlycoLink and CarboxyLink Gels?

GlycoLink gel is hydrazide-activated crosslinked beaded agarose, and it is useful for coupling glycoproteins via aldehydes formed from their sugars by sodium meta-periodate oxidation. Reaction of aldehydes with hydrazide- activated resin is catalyzed by aniline resulting in >90% coupling in 4 hours or less. CarboxyLink gel on the other hand is crosslinked beaded agarose activated with diaminodipropylamine (DADPA) and is useful for immobilizing carboxyl- containing biomolecules after EDC activation. Actually, both resins can be used with EDC to couple ligands via carboxylic acids.

Note: Both immobilization chemistries are available on UltraLink Support as UltraLink Hydrazide and UltraLink DADPA respectively.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.