ClipTip™ Filtered Pipette Tips, Racked, Sterile
ClipTip™ Filtered Pipette Tips, Racked, Sterile
Actual product may vary
ClipTip™ Filtered Pipette Tips, Racked, Sterile
ClipTip™ Filtered Pipette Tips, Racked, Sterile
ClipTip™ Filtered Pipette Tips, Racked, Sterile
ClipTip™ Filtered Pipette Tips, Racked, Sterile
Thermo Scientific™

ClipTip™ Filtered Pipette Tips, Racked, Sterile

Ensure a secure seal for every sample. A unique, innovative interlocking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
94420313-HID200 μL
94420063-HID12.5 μL
94420213-HID20 μL
94420713-HID1000 μL
Catalog number 94420313-HID
Price (USD)
138.00
Pack of 960
Add to cart
Volume (Metric):
200 μL
Price (USD)
138.00
Pack of 960
Add to cart
Ensure a secure seal for every sample. Thermo Scientific™ ClipTip™ Pipette Tips provide a secure seal for every sample with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force. Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip and E1-ClipTip pipettes until — and only until — it is released.

Key Features
• ClipTip pipette tips feature 'clips' that lock tips in place and form a complete seal
• Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure
• ClipTip pipette tips are clipped securely on the F1-ClipTip and E1-ClipTip pipettes with a light touch eliminating the frustration of banging tips on your pipette
• Filtered ClipTip pipette tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations

Low retention
Low retention technology minimizes liquid retention, and enable maximium sample recovery. Available in ClipTip 20-1250 96-format tips.

Proven quality
Certified free of RNase, DNase, DNA, ATP and endotoxin contamination

Environmentally responsible
ClipTip reload system uses recycled components and 30% less plastic

Versatile Tip Range
• 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
• 384-format volumes: 12.5, 30 and 125 μL
• Extended lenght tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables

Immediate traceability
Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert

Note:
Availability may vary by country

Specifications
DescriptionClipTip 200, Filtered, Sterile, Rack
Volume (Metric)200 μL
FilteredFiltered
SterilitySterile
AutoclavableNot Autoclavable
Length (English)2.2 in.
Length (Metric)5.6 cm
FormatRack
Tip StyleLow Retention
Quantity10 Racks (96 Tips/Rack)
Color CodeYellow
For Use With (Equipment)Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip
Product LineClipTip
Fit for PurposeForensic Ready
Unit SizePack of 960