Methanol, UHPLC-MS, Thermo Scientific™
This product ships via ground from US northeast.
Methanol, UHPLC-MS, Thermo Scientific™
Methanol, UHPLC-MS, Thermo Scientific™
Methanol, UHPLC-MS, Thermo Scientific™
Methanol, UHPLC-MS, Thermo Scientific™
Thermo Scientific™

Methanol, UHPLC-MS, Thermo Scientific™

Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer.
Have Questions?
Catalog NumberQuantityPackaging
A4581CS
also known as A4581
1 LClear Borosilicate Glass Bottle
Catalog number A4581CS
also known as A4581
Price (USD)
684.00
Case of 6
Add to cart
Quantity:
1 L
Packaging:
Clear Borosilicate Glass Bottle
Price (USD)
684.00
Case of 6
Add to cart
Methanol, CH4O, CAS Number-67-56-1, wood naphtha, pyroxylic spirit, methyl alcohol, methylol, columbian spirit, colonial spirit, wood spirit, wood alcohol, methyl hydroxide, carbinol, 1L
RUO – Research Use Only
Specifications
Chemical Name or MaterialMethanol
Melting Point-98°C
Absorbance≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.4 AU at 215nm, ≤000 5 AU at 260nm, ≤02. AU at 220nm, ≤05. AU at 210nm
CAS67-56-1
ColorColorless
Recommended StorageRoom temperature
Density0.791 g/cm3
Boiling Point64.7°C
Quantity1 L
PackagingClear Borosilicate Glass Bottle
Physical FormLiquid
Assay Percent Range99.9% minimum by GC
Residue after Evaporation1ppm max.
Molecular FormulaCH4O
Titratable Acid0.0003meq/g max.
Linear FormulaCH3OH
Titratable Base0.0002meq/g max.
MDL NumberMFCD00004595
Vapor Pressure128 hPa at 20°C
Viscosity0.55 cP at 20°C
Water0.02% Max.
Synonymmethyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
IUPAC Namemethanol
Molecular Weight (g/mol)32.04
PubChem CID887
ChEBICHEBI:17790
GradeUHPLC-MS
Unit SizeCase of 6