Platinum™ SuperFi™ U Multiplex Master Mix
This product will be discontinued on April 30th, 2025. If you have any questions, please contact Customer Support.
Platinum™ SuperFi™ U Multiplex Master Mix
Invitrogen™

Platinum™ SuperFi™ U Multiplex Master Mix

Invitrogen Platinum SuperFi U Multiplex Master Mix is a product designed to synthesize multiple custom amplicons for further use inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Samples
A514009696 preps
A514002424 preps
Catalog number A5140096
Price (USD)
356.00
Each
Add to cart
No. of Samples:
96 preps
Price (USD)
356.00
Each
Add to cart
Invitrogen Platinum SuperFi U Multiplex Master Mix is a product designed to synthesize multiple custom amplicons for further use in next-generation sequencing. The ready-to-use 4X master mix contains uracil-tolerant high-fidelity DNA polymerase with all necessary buffer components and dNTPs for efficient targeted amplification.

Features of Platinum SuperFi U Multiplex Master Mix include:
• Optimized for high multiplex PCR
• Enables fast, efficient and robust amplification
• Provides high yields

Specifications
ColorTransparent
Concentration4X
Sample TypegDNA, cDNA
EnzymePlatinum™ SuperFi™ U DNA Polymerase
For Use With (Application)Sars-Covid-2 Sequencing
No. of Reactions96 Preps
No. of Samples96 preps
Product LineCollibri™
Product TypeMaster Mix
SpeciesAll
Unit SizeEach
Contents & Storage
Store at -25°C to -15°C