Sensititre™ Streptococcus FDANDSF AST Plate
Sensititre™ Streptococcus FDANDSF AST Plate
Thermo Scientific™
Sensititre™ Streptococcus FDANDSF AST Plate
Accurately perform antimicrobial susceptibility testing (AST) Streptococcus species using this FDA-cleared, convenient plate.
Have Questions?
Catalog NumberVolume Per Well
FDANDSF100 μL
Catalog number FDANDSF
Price (USD)
-
Request A Quote
Volume Per Well:
100 μL
Accurately perform antimicrobial susceptibility testing (AST) of Streptococcus species using the FDA-cleared Thermo Scientific™ Sensititre™ Streptococcus FDANDSF AST Plate with Oritavancin, Dalbavancin, and Tedizolid. Obtain true minimum inhibitory concentration (MIC) results, versus extrapolated MIC results. The addition of antimicrobials, oritavancin, dalbavancin, and tedizolid, offers more susceptibility testing options and aids targeted treatment decisions for serious skin infections.
Expand treatment options for serious skin infections. Perform oritavancin, dalbavancin, and tedizolid susceptibility testing of fastidious Gram-positive organisms that cause acute bacterial skin and skin structure infections (ABSSSIs) in adults using Sensititre Streptococcus FDANDSF AST Plate with Oritavancin, Dalbavancin, and Tedizolid.
  • Robust: accurate results the first time, thereby reducing unnecessary testing and protocols
  • Easy to store: room temperature storage relieves cold storage restrictions, simplifying inventory control
  • Economic: individual packaging allows you to test one plate at a time with no waste
  • Flexible: automated and manual read options to suit your workflow
The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
  • Scalable: flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
  • Comprehensive: over 240 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
  • Tailor-made: design your own AST panel to meet specific formulary requirements and prescription protocols and to provide clinicians results that are based on real-time trends and local epidemiology
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
0.5 McFarland StandardMHB 5mL (Part no. T3462-05)
SpeciesStreptococcus pneumoniae, Streptococcus species
Antimicrobial AgentsVarious
Broth TypeMHB w/LHB (Part no. CP114)
DescriptionSensititre Streptococcus Plate with Oritavancin, Dalbavancin, Tedizolid
Antibiotic CodeVarious
Product TypeAST Plate
Test TypeMinimum Inhibitory Concentration (MIC)
For Use With (Application)Streptococcus pyogenes, Streptococcus agalactiae, Streptococcus dysgalactia, Streptococcus anginosus
Incubation Conditions34-36°C; O2
Incubation Time20-24 hours
Isolates Per Plate1
Packaging TypeBox
Quantity10/Box
Reading OptionsAutoread or manual
Transfer Volume100µL
Volume Per Well100 μL
Unit SizeEach