Cyto-Plex™ Carboxylated Particles
Cyto-Plex™ Carboxylated Particles
Actual product may vary
Cyto-Plex™ Carboxylated Particles
Cyto-Plex™ Carboxylated Particles
Cyto-Plex™ Carboxylated Particles
Cyto-Plex™ Carboxylated Particles
Thermo Scientific™

Cyto-Plex™ Carboxylated Particles

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDiameter (Metric)QuantityLevel
FM4CR05Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 5
FM4CR01Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 1
FM4CR01BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 1
FM4CR02Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 2
FM4CR02BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 2
FM4CR03Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 3
FM4CR03BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 3
FM4CR04Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 4
FM4CR04BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 4
FM4CR05BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 5
FM4CR06Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 6
FM4CR06BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 6
FM4CR07Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 7
FM4CR07BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 7
FM4CR08Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 8
FM4CR08BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 8
FM4CR09Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 9
FM4CR09BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 9
FM4CR10Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 10
FM4CR10BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 10
FM4CR11Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 11
FM4CR11BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 11
FM4CR12Cyto-Plex Carboxylated Particles, 4µm4 μm1 mLLevel 12
FM4CR12BCyto-Plex Carboxylated Particles, 4µm4 μm5 mLLevel 12
FM5CR01Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 1
FM5CR01BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 1
FM5CR02Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 2
FM5CR02BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 2
FM5CR03Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 3
FM5CR03BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 3
FM5CR04Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 4
FM5CR04BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 4
FM5CR05Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 5
FM5CR05BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 5
FM5CR06Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 6
FM5CR06BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 6
FM5CR07Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 7
FM5CR07BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 7
FM5CR08Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 8
FM5CR08BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 8
FM5CR09Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 9
FM5CR09BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 9
FM5CR10Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 10
FM5CR10BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 10
FM5CR11Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 11
FM5CR11BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 11
FM5CR12Cyto-Plex Carboxylated Particles, 5µm5 μm1 mLLevel 12
FM5CR12BCyto-Plex Carboxylated Particles, 5µm5 μm5 mLLevel 12
Catalog number FM4CR05
Price (USD)
-
Request A Quote
Description:
Cyto-Plex Carboxylated Particles, 4µm
Diameter (Metric):
4 μm
Quantity:
1 mL
Level:
Level 5
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.
  • Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
  • Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
  • Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
  • High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
  • Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
  • Emission can be collected in either the PE-Cy5 or APC channels
  • Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters
  • Open platform microspheres for suspension array analysis in flow cytometry
  • Specifications
    AdditiveSodium Azide Preservative, 0.05% Tween-20
    ContentsPolystyrene microspheres in water
    DescriptionCyto-Plex Carboxylated Particles, 4µm
    ColorRed
    MaterialPolystyrene with Copolymer Grafted Surface
    Product TypeCarboxylated Particle
    Dye TypeFirefli™ Fluorescent Red (635/700nm)
    IncludesMaterial Safety Data Sheet (MSDS) and Package Insert Sheet
    LevelLevel 5
    Particle Density1.06g/cm³
    Quantity1 mL
    StabilityProduct stable for a minimum of 12 months
    Surface ModificationCarboxylate-Modified
    Uniformity≤2% CV
    Diameter (Metric)4 μm
    Unit SizeEach
    Contents & Storage
    Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer.