GeneBLAzer™ PAC1-CRE-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ PAC1-CRE-bla CHO-K1 Cells

The GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells contain the human Adenylate Cyclase Activating Polypeptide 1 Receptor 1 (ADCYAP1R/PAC1) stably integrated into theRead more
Have Questions?
Catalog number K1525
Price (USD)
10,930.00
Each
Add to cart
Price (USD)
10,930.00
Each
Add to cart
The GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells contain the human Adenylate Cyclase Activating Polypeptide 1 Receptor 1 (ADCYAP1R/PAC1) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat #K1129) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element (CRE). The PAC1-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of established ligands. In addition, PAC1-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_001118
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
Gene SymbolADCYAP1R1
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryADCYAP1R1, PAC1
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit SizeEach
Contents & Storage
K1525 GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells
• GeneBLAzer™ PAC1-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.