GeneBLAzer™ CCKAR HEK 293T DA Assay Kit
Product Image
Thermo Scientific™

GeneBLAzer™ CCKAR HEK 293T DA Assay Kit

The GeneBLAzer™ CCKAR-NFAT-bla CHO-K1 cells contain the human Cholecystokinin A Receptor (CCKAR) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cellRead more
Have Questions?
Catalog number K1623
Price (USD)
1,182.00
Each
Add to cart
Price (USD)
1,182.00
Each
Add to cart

The GeneBLAzer™ CCKAR-NFAT-bla CHO-K1 cells contain the human Cholecystokinin A Receptor (CCKAR) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line. GeneBLAzer™ NFAT-bla CHO-K1 (Cat. No. K1447) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element stably integrated into CHO-K1 cells.

The CCKAR-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations using established ligands. In addition, CCKAR-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary. Division Arrested cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000730.2
Assay EntryCell-based beta lactamase reporter gene
Cell StateDivision-Arrested Cells
Detection MethodFluorescence
Gene SymbolCCKAR
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity8 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryCCKAR
Cell Line293T (HEK)
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit SizeEach
Contents & Storage
Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol).

Includes:
• GeneBLAzer™ CCKAR HEK 293T DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D
• GeneBLAzer™ CCKAR HEK 293T DA Assay Protocol
• LiveBLAzer™-FRET B/G Loading Protocol
• Certificate of Analysis

The GeneBLAzer™ CCKAR HEK 293T DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.