CellSensor™ SIE-bla ME-180 Cell Line
Product Image
Thermo Scientific™

CellSensor™ SIE-bla ME-180 Cell Line

The CellSensor&tarde; SIE-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the Sis-Inducible Element (SIE) stably integratedRead more
Have Questions?
Catalog number K1648
Price (USD)
10,930.00
Each
Add to cart
Price (USD)
10,930.00
Each
Add to cart

The CellSensor&tarde; SIE-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the Sis-Inducible Element (SIE) stably integrated into ME-180 cells. To construct this cell line, the SIE-bla construct was transduced into ME-180 cells by lentivirus. Flow cytometry was used to isolate cells responsive to Interleukin-6 (IL-6). This cell line is validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z' and EC50 concentrations of IL-6. The CellSensor™ SIE-bla ME-180 cell line is responsive to IL-6, OSM and IFNγ and can be used to probe the JAK/STAT signaling pathway. This cell line can be adapted for high-throughput screening of agonist or antagonist compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Product LineCellSensor™
Quantity1 Vial
Target EntrySTAT1, STAT3, JAK/STAT (IFN gamma, IL-6/OSM) pathway, JAK2, JAK1
System TypeCellSensor™
Druggable TargetSignaling Pathway
Unit SizeEach
Contents & Storage
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.