GeneBLAzer™ M4-Gqo5-NFAT-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ M4-Gqo5-NFAT-bla CHO-K1 Cells

GeneBLAzer™ M4 Gqo5-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 4 receptor (M4) stably integrated into the GeneBLAzer™ Gqo5-NFAT-blaRead more
Have Questions?
Catalog number K1741
Price (USD)
10,930.00
Each
Add to cart
Price (USD)
10,930.00
Each
Add to cart

GeneBLAzer™ M4 Gqo5-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 4 receptor (M4) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells (Cat. No. K1536) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Chimeric G protein, Gqo5.

The GeneBLAzer™ M4 Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Cabachol. In addition, GeneBLAzer™ M4 Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The clinical implications of the M4 receptor are unknown, however studies with knockout mice suggest that M4 may have implications in dopaminergic function, karatinocyte migration and wound healing, anxiolysis, and analgesia. In addition, M4 may be important in memory circuits and have implications in Alzheimer's Disease. Additional information on the muscarinic receptors can be found in literature.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000741
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
For Use With (Application)FRET
Gene SymbolM4
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryM4
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit SizeEach
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ M4 Gqo5 CHO-K1 DA cells
• GeneBLAzer™ M4 Gqo5 CHO-K1 DA Assay Protocol
• Certificate of Analysis
• Solution D
• LiveBLAzer™-FRET B/G Loading Protocol

The GeneBLAzer™ M4-Gqo5-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.