Spectra™ Strep A Agar
Spectra™ Strep A Agar
Spectra™ Strep A Agar
Spectra™ Strep A Agar
Spectra™ Strep A Agar
Spectra™ Strep A Agar
Thermo Scientific™

Spectra™ Strep A Agar

Detect and identify group A streptococci (GAS) with this chromogenic medium.
Have Questions?
Catalog number R01812
Price (USD)
-
Request A Quote
Isolate and differentiate group A streptococci (GAS) colonies among other bacteria in the complex throat flora with easy to read, chromogenic Thermo Scientific™ Spectra™ Strep A Agar.

Streptococcus pyogenes infection is responsible for nearly all Streptococci caused bacterial pharyngitis, which can also lead to Scarlett fever and Rheumatic fever in children. This is seen most commonly among children 5 through 15 years of age. The use of antibiotics is recommended to treat GAS pharyngitis in children to shorten the duration of symptoms, reduce the likelihood of transmission to others and prevent the development of complications, including acute rheumatic fever, peritonsillar abscess, and mastoiditis.

Rapid and accurate detection of this microorganism is essential to start antibiotic therapy,  prevent complications, and reduce the risk of transmission to other persons.

Traditional culture methods provide poor sensitivity for isolating Streptococcus pyogenes. The throat culture is used primarily as a confirmatory test with definitive results requiring 48 hours.

The new chromogenic formulation Thermo Scientific Spectra Strep A Agar allows an easy-to-read and straightforward color differentiation of GAS colonies (in orange to red) among the other bacteria in the complex throat flora (colorless or blue colonies).

Easy to read:

  • Target organism in orange to red
  • Other non-target colorless or blue colonies

 

Fast time to results

  • Reduce reporting time to 24 hours

 

Compatibility with lab automation systems

  • Provides efficient and consistent processi
  • Imaging allows for high-resolution digital imaging analysis.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
ColorLight Amber
DescriptionStrep A Chrmogenic Agar
Format85 mm monoplate
Manufacturing OriginUnited States
Product TypeAgar
Packaging TypeFlow wrapped - single layer
Quantity10/Pk.
Unit SizeEach
Contents & Storage
2°C to 8°C