Savant™ SpeedVac™ Refrigerated Vapor Trap Series
Product Image
Thermo Scientific™
Savant™ SpeedVac™ Refrigerated Vapor Trap Series
Condense and trap a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionElectrical Requirements
RVT450-115Refrigerated Vapor Cold Trap RVT450115 V 60 Hz
RVT450-230Refrigerated Vapor Cold Trap RVT450230 V 50 Hz
Catalog number RVT450-115
Price (USD)
-
Request A Quote
Description:
Refrigerated Vapor Cold Trap RVT450
Electrical Requirements:
115 V 60 Hz
Condenses and removes a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps (RVT). The RVT safely recovers the solvent and reduces emissions of hazardous chemicals in the environment.
Solvents are removed from samples during vacuum evaporation with a SpeedVac™ Concentrator which are then collected in a CFC-free Refrigerated Vapor Trap (RVT).   Solvent vapors pass into a Glass Condensation Flask and condense on its walls. This prevents the vapors from reaching and potentially damaging the oil-sealed vacuum pump. 
  • Rapid cooling time minimizes waiting period
  • Easy to remove glass flask for quick solvent changes
  • Rapidly cools to operating temperature
  • Easy to use glass condensation flask allows for the collection of liquid into a self-contained jar for easy draining
  • Sealed refrigeration system eliminates consumable coolants such as dry ice and acetone
Specifications
Certifications/ComplianceUL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
DescriptionRefrigerated Vapor Cold Trap RVT450
TypeRefrigerated Vapor Trap
Electrical Requirements115 V 60 Hz
Unit SizeEach