Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G18830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332373.1 | NP_001320897.1 | 11 - 12 | 1198 | 162 | |
NM_001332374.1 | NP_001320898.1 | 11 - 12 | 1163 | 162 | ||
NM_101740.3 | NP_173317.2 | 11 - 12 | 1140 | 162 |
Set Membership: |
![]() ![]() ![]() ![]() |