Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNF7.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335643.1 | NP_001318249.1 | 4 - 5 | 407 | 68 | |
NM_127541.5 | NP_179573.1 | 6 - 7 | 679 | 68 | ||
GenBank mRNA | AY045911.1 | - | 6 - 7 | 591 | 68 | |
AY079421.1 | - | 5 - 6 | 460 | 68 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |