Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRK2.9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127867.5 | NP_179885.1 | 6 - 7 | 667 | 83 |
Set Membership: |
![]() ![]() ![]() ![]() |