Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202710.1 | NP_001189639.1 | 7 - 8 | 1170 | 133 | |
NM_128592.3 | NP_180598.1 | 7 - 8 | 1174 | 133 | ||
GenBank mRNA | BT000465.1 | - | 7 - 8 | 1117 | 133 | |
BT008877.1 | - | 7 - 8 | 1011 | 133 |
Set Membership: |
![]() ![]() ![]() ![]() |