Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124929.3 | NP_200358.1 | 9 - 10 | 1026 | 127 | |
GenBank mRNA | AK221571.1 | - | 9 - 10 | 1026 | 127 |
Set Membership: |
![]() ![]() ![]() ![]() |