Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119572.7 | NP_195137.5 | 3 - 4 | 1485 | 148 | |
GenBank mRNA | BT002354.1 | - | 3 - 4 | 1044 | 148 | |
L19418.1 | - | 3 - 4 | 1375 | 148 |
Set Membership: |
![]() ![]() ![]() ![]() |