Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100276.4 | NP_563696.1 | 4 - 5 | 321 | 122 | |
GenBank mRNA | AY052238.1 | - | 4 - 5 | 316 | 122 | |
AY060501.1 | - | 4 - 5 | 168 | 122 |
Set Membership: |
![]() ![]() ![]() ![]() |