Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS24.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122201.2 | NP_197686.1 | 4 - 5 | 556 | 80 | |
GenBank mRNA | AY054614.1 | - | 4 - 5 | 488 | 80 | |
AY128742.1 | - | 4 - 5 | 410 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() |