Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105415.1 | NP_001098885.1 | 8 - 9 | 1302 | 65 | |
GenBank mRNA | BC134486.1 | - | 8 - 9 | 1302 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |