Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Bt03215936_m1
          See other PSEN1 GE Assays ›
          Gene Symbol:
          PSEN1
          Gene Name:
          presenilin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 84619595 - 84686880 on Build ARS-UCD1.2
          Species:
          Bovine
          Species Specific ID (Flybase ID):
          -
          Assay ID Bt03215936_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PSEN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_174721.2 NP_777146.1 4 - 5 558 95
          GenBank mRNA AF038936.1 - 4 - 5 558 95
          BC151458.1 - 6 - 7 759 95

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          negative regulation of transcription by RNA polymerase II
          blood vessel development
          cell fate specification
          somitogenesis
          neuron migration
          positive regulation of receptor recycling
          heart looping
          positive regulation of L-glutamate import across plasma membrane
          hematopoietic progenitor cell differentiation
          astrocyte activation involved in immune response
          T cell activation involved in immune response
          protein glycosylation
          membrane protein ectodomain proteolysis
          mitochondrial transport
          cellular response to DNA damage stimulus
          response to oxidative stress
          negative regulation of epidermal growth factor-activated receptor activity
          Notch signaling pathway
          memory
          post-embryonic development
          regulation of neuron projection development
          protein transport
          choline transport
          synaptic vesicle targeting
          protein processing
          cerebral cortex cell migration
          Cajal-Retzius cell differentiation
          dorsal/ventral neural tube patterning
          embryonic limb morphogenesis
          positive regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          endoplasmic reticulum calcium ion homeostasis
          amyloid-beta formation
          intracellular signal transduction
          positive regulation of protein import into nucleus
          amyloid precursor protein metabolic process
          myeloid dendritic cell differentiation
          positive regulation of apoptotic process
          positive regulation of catalytic activity
          skin morphogenesis
          positive regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          regulation of synaptic plasticity
          thymus development
          neuron development
          skeletal system morphogenesis
          brain morphogenesis
          epithelial cell proliferation
          negative regulation of axonogenesis
          synapse organization
          positive regulation of coagulation
          T cell receptor signaling pathway
          neuron apoptotic process
          negative regulation of ubiquitin-protein transferase activity
          regulation of synaptic transmission, glutamatergic
          regulation of resting membrane potential
          regulation of canonical Wnt signaling pathway
          positive regulation of dendritic spine development
          calcium ion transmembrane transport
          modulation of age-related behavioral decline
          cell-cell adhesion
          cellular response to amyloid-beta
          negative regulation of core promoter binding
          positive regulation of amyloid fibril formation
          neuron projection maintenance
          negative regulation of ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic signaling pathway
          calcium channel activity
          PDZ domain binding
          aspartic endopeptidase activity, intramembrane cleaving

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0