Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7L3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078161.2 | NP_001071629.1 | 1 | 709 | 79 | |
GenBank mRNA | BC126777.1 | - | 1 | 648 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |