Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083519.1 | NP_001076988.1 | 1 - 2 | 495 | 71 | |
GenBank mRNA | BC123676.1 | - | 1 - 2 | 495 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |