Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110068.2 | NP_001103538.1 | 3 - 4 | 298 | 121 | |
GenBank mRNA | BC108159.1 | - | 1 | 299 | 121 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |