Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078221.7 | NP_510622.3 | 11 - 12 | 1456 | 139 |
Set Membership: |
![]() ![]() ![]() ![]() |