Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ech-1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059388.8 | NP_491789.1 | 4 - 5 | 1074 | 95 | |
NM_059389.5 | NP_491790.1 | 10 - 11 | 2507 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() |