Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059090.9 | NP_491491.2 | 7 - 8 | 1486 | 157 |
Set Membership: |
![]() ![]() ![]() ![]() |