Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027043.3 | NP_001022214.1 | 6 - 7 | 1373 | 95 | |
NM_001027044.4 | NP_001022215.1 | 6 - 7 | 1377 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() |