Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063213.7 | NP_495614.4 | 2 - 3 | 238 | 105 | |
GenBank mRNA | AF497824.1 | - | 1 - 2 | 218 | 105 |
Set Membership: |
![]() ![]() ![]() ![]() |