Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srz-70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067954.2 | NP_500355.2 | 3 - 4 | 524 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |