Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc-73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068064.7 | NP_500465.4 | 1 - 2 | 73 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() |