Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyc-2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383085.1 | 2 - 3 | 509 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |