Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lipl-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072354.4 | NP_504755.3 | 7 - 8 | 1418 | 154 |
Set Membership: |
![]() ![]() ![]() ![]() |